Assay Details
Target Gene Details
Entrez Gene ID: | 246243 |
Gene Name: | ribonuclease H1 |
Gene Aliases: |
H1RNA, PEOB2, RNH1 |
Location: |
Chr.2:3531813-3558371 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASEH1 | NM_001286834.1 | 8 | 1557 | NP_001273763.1 |
| NM_001286837.1 | 8 | 1795 | NP_001273766.1 | |
| NM_002936.4 | 8 | 1557 | NP_002927.2 | |
| AK075490.1 | 8 | 1506 | ||
| AK096913.1 | 8 | 1713 | ||
| DA612116.1 | 1 | 262 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011656 | Chr.2:2931099 - 3576389 on Build GRCh38 | Gain |
|
| dgv3702n100 | Chr.2:3361936 - 3733306 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map