Assay Details
Target Gene Details
Entrez Gene ID: | 7158 |
Gene Name: | tumor protein p53 binding protein 1 |
Gene Aliases: |
53BP1, TDRD30, TP53, p202, p53BP1 |
Location: |
Chr.15:43403064-43510509 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TP53BP1 | NM_001141979.1 | 24 | 5255 | NP_001135451.1 |
| NM_001141980.1 | 24 | 5261 | NP_001135452.1 | |
| NM_005657.2 | 24 | 5281 | NP_005648.1 | |
| XM_011521985.1 | 23 | 5110 | XP_011520287.1 | |
| XM_017022533.1 | 25 | 5365 | XP_016878022.1 | |
| AB210025.1 | 24 | 5226 | ||
| AF078776.1 | 24 | 5306 | AAC62018.1 | |
| AK123418.1 | 3 | 1267 | ||
| BC112161.1 | 24 | 5241 | AAI12162.1 | |
| BX537418.1 | 24 | 5260 | CAD97660.1 | |
| U09477.1 | 11 | 2300 | AAA21596.1 |
More Information
Set Membership: |
|

Genomic Map