Assay Details
Target Gene Details
Entrez Gene ID: | 474354 |
Gene Name: | leucine rich repeat containing 18 |
Gene Aliases: |
UNQ933, UNQ9338, VKGE9338 |
Location: |
Chr.10:48881011-48940126 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC18 | NM_001006939.3 | NP_001006940.3 | ||
| XM_006717867.2 | XP_006717930.1 | |||
| XM_011539822.1 | XP_011538124.1 | |||
| XM_011539823.1 | XP_011538125.1 | |||
| XM_011539826.2 | XP_011538128.1 | |||
| XM_011539828.2 | XP_011538130.1 | |||
| XM_011539829.2 | XP_011538131.1 | |||
| XM_017016276.1 | XP_016871765.1 | |||
| XM_017016277.1 | XP_016871766.1 | |||
| AY358137.1 | 2 | 825 | AAQ88504.1 | |
| BC036722.1 | AAH36722.1 |
Target Gene Details
Entrez Gene ID: | 57705 |
Gene Name: | WDFY family member 4 |
Gene Aliases: |
C10orf64 |
Location: |
Chr.10:48684815-48982956 on Build GRCh38 |
Assay Gene Location: | Within Intron 53 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDFY4 | NM_020945.1 | NP_065996.1 | ||
| XM_005270004.3 | XP_005270061.1 | |||
| XM_011539986.2 | XP_011538288.2 | |||
| XM_011539987.2 | XP_011538289.2 | |||
| XM_011539988.2 | XP_011538290.1 | |||
| XM_017016463.1 | XP_016871952.1 | |||
| XM_017016464.1 | XP_016871953.1 | |||
| AB046827.1 | BAB13433.1 | |||
| AK074085.1 | BAB84911.1 | |||
| AK074443.1 | ||||
| AL833669.1 | ||||
| BC047574.1 | AAH47574.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831865 | Chr.10:48786209 - 48956194 on Build GRCh38 | Gain |
|
| nsv1042677 | Chr.10:47993784 - 49928329 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map