Assay Details
Target Gene Details
Entrez Gene ID: | 344838 |
Gene Name: | progestin and adipoQ receptor family member 9 |
Gene Aliases: |
- |
Location: |
Chr.3:142961231-142963336 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAQR9 | NM_198504.2 | 1 | 878 | NP_940906.1 |
| AK093381.1 | 1 | 99 | ||
| AK123932.1 | 1 | 878 | BAC85729.1 | |
| AY424287.1 | 1 | 955 | AAR08375.1 | |
| BC118666.1 | 1 | 878 | ||
| BC122527.1 | 1 | 878 |
More Information
Set Membership: |
|

Genomic Map