Assay Details
Target Gene Details
Entrez Gene ID: | 4308 |
Gene Name: | transient receptor potential cation channel subfamily M member 1 |
Gene Aliases: |
CSNB1C, LTRPC1, MLSN1 |
Location: |
Chr.15:31001061-31161273 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPM1 | NM_001252020.1 | 27 | 4370 | NP_001238949.1 |
| NM_001252024.1 | 28 | 4158 | NP_001238953.1 | |
| NM_002420.5 | 27 | 4072 | NP_002411.3 | |
| AB115498.1 | 24 | 3729 | BAC80200.1 | |
| AB115499.1 | 24 | 3729 | BAC80201.1 | |
| AB115500.1 | 23 | 3584 | BAC80202.1 | |
| AB115501.1 | 24 | 3729 | BAC80203.1 | |
| AB115502.1 | 24 | 3711 | BAC80204.1 | |
| AF071787.1 | 27 | 4049 | AAC80000.1 | |
| HM135790.1 | 28 | 4153 | ||
| HM135791.1 | 27 | 4370 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832951 | Chr.15:30839036 - 31012266 on Build GRCh38 | Loss |
|
| nsv525795 | Chr.15:30644082 - 31032328 on Build GRCh38 | Gain |
|
| esv33507 | Chr.15:30891121 - 31178820 on Build GRCh38 | Loss |
|
| esv3636036 | Chr.15:30818302 - 31038636 on Build GRCh38 | Gain |
|
| nsv832952 | Chr.15:30912223 - 31041795 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map