Assay Details
Target Gene Details
Entrez Gene ID: | 55337 |
Gene Name: | chromosome 19 open reading frame 66 |
Gene Aliases: |
RyDEN |
Location: |
Chr.19:10086119-10093252 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C19orf66 | NM_001308277.1 | NP_001295206.1 | ||
| NM_018381.3 | NP_060851.2 | |||
| XM_011528121.1 | XP_011526423.1 | |||
| XM_017026934.1 | XP_016882423.1 | |||
| AK002148.1 | BAA92109.1 | |||
| AK096142.1 | BAC04710.1 | |||
| AL137694.1 | ||||
| BC035817.2 | AAH35817.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2414 | Chr.19:10063769 - 10097402 on Build GRCh38 | Insertion |
|
| nsv458355 | Chr.19:10061840 - 10102478 on Build GRCh38 | Loss |
|
| nsv833743 | Chr.19:10077649 - 10251913 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map