Assay Details
Target Gene Details
Entrez Gene ID: | 56903 |
Gene Name: | poly(A) polymerase beta |
Gene Aliases: |
PAPT, TPAP |
Location: |
Chr.7:4857738-4861994 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAPOLB | NM_020144.4 | 1 | 876 | NP_064529.4 |
| AF218840.1 | 1 | 797 | AAF81013.1 | |
| AK312788.1 | 1 | 907 | ||
| BC036653.2 | 1 | 876 | AAH36653.2 |
Target Gene Details
Entrez Gene ID: | 55698 |
Gene Name: | Rap associating with DIL domain |
Gene Aliases: |
RASIP2 |
Location: |
Chr.7:4799109-4883704 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RADIL | NM_018059.4 | NP_060529.4 | ||
| AB058752.1 | ||||
| AK294745.1 | ||||
| BC117317.1 | ||||
| BC126311.1 | ||||
| BC143526.1 | ||||
| EF560723.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv605980 | Chr.7:4836426 - 4963742 on Build GRCh38 | Loss |
|
| nsv605979 | Chr.7:4814961 - 4904883 on Build GRCh38 | Loss |
|
| esv3611950 | Chr.7:4828936 - 4874508 on Build GRCh38 | Loss |
|
| nsv1029123 | Chr.7:4782103 - 4879646 on Build GRCh38 | Loss |
|
| dgv6238n100 | Chr.7:4810816 - 5030133 on Build GRCh38 | Loss |
|
| nsv528748 | Chr.7:4780950 - 4971362 on Build GRCh38 | Loss |
|
| nsv8033 | Chr.7:4762656 - 5046966 on Build GRCh38 | Loss |
|
| nsv823994 | Chr.7:4752754 - 4888697 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map