Assay Details
Target Gene Details
Entrez Gene ID: | 84939 |
Gene Name: | melanoma associated antigen (mutated) 1 |
Gene Aliases: |
EXPAND1, HSPC211, MUM-1 |
Location: |
Chr.19:1354977-1378431 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MUM1 | NM_032853.3 | NP_116242.2 | ||
| NR_024247.1 | ||||
| XM_011528380.2 | XP_011526682.1 | |||
| XM_011528382.2 | XP_011526684.1 | |||
| XM_011528383.1 | XP_011526685.1 | |||
| XM_011528384.2 | XP_011526686.1 | |||
| XM_011528385.2 | XP_011526687.1 | |||
| XM_017027401.1 | XP_016882890.1 | |||
| AK309092.1 | ||||
| AK310721.1 | ||||
| BC110874.1 | AAI10875.1 | |||
| BC130443.1 | ||||
| BC144138.1 | ||||
| BI553935.1 | ||||
| BQ895780.1 | ||||
| BU541563.1 | ||||
| DA739708.1 | ||||
| DN993266.1 | ||||
| U20896.1 | AAC50239.1 | |||
| U20897.1 | AAC50240.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055164 | Chr.19:1246647 - 1389804 on Build GRCh38 | Gain |
|
| esv32942 | Chr.19:1320268 - 1999205 on Build GRCh38 | Gain+Loss |
|
| esv3643389 | Chr.19:1264454 - 1397379 on Build GRCh38 | Gain |
|
| esv2758743 | Chr.19:951641 - 1544934 on Build GRCh38 | Gain+Loss |
|
| esv3643396 | Chr.19:1356856 - 1396468 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map