Assay Details
Target Gene Details
Entrez Gene ID: | 80314 |
Gene Name: | enhancer of polycomb homolog 1 |
Gene Aliases: |
Epl1 |
Location: |
Chr.10:32267716-32378798 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPC1 | NM_001272004.1 | 10 | 1931 | NP_001258933.1 |
| NM_001272019.2 | 10 | 1537 | NP_001258948.1 | |
| NM_001282391.1 | 10 | 1783 | NP_001269320.1 | |
| NM_025209.3 | 10 | 1931 | NP_079485.1 | |
| XM_006717510.1 | 10 | 1783 | XP_006717573.1 | |
| AF277374.1 | 10 | 1898 | AAG41402.1 | |
| AF286905.1 | 10 | 1601 | AAK60501.1 | |
| AK024329.1 | 7 | 1046 | BAB14888.1 | |
| AK092304.1 | 10 | 1537 | BAC03857.1 | |
| AK299676.1 | 10 | 1724 | ||
| AK309309.1 | 10 | 1669 | ||
| AK309822.1 | 10 | 1800 | ||
| BC036529.1 | 10 | 1932 | AAH36529.1 | |
| CR749375.1 | 8 | 4524 | CAH18228.1 |
More Information
Set Membership: |
|

Genomic Map