Assay Details
Target Gene Details
Entrez Gene ID: | 4690 |
Gene Name: | NCK adaptor protein 1 |
Gene Aliases: |
NCK, NCKalpha, nck-1 |
Location: |
Chr.3:136862208-136951610 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCK1 | NM_001291999.1 | NP_001278928.1 | ||
| NM_006153.5 | NP_006144.1 | |||
| AK312692.1 | ||||
| BC006403.2 | AAH06403.1 | |||
| DA265476.1 | ||||
| DA855124.1 | ||||
| DB117533.1 | ||||
| KU178218.1 | ||||
| KU178219.1 | ||||
| KU178220.1 | ||||
| X17576.1 | CAA35599.1 |
More Information
Set Membership: |
|

Genomic Map