Assay Details
Target Gene Details
Entrez Gene ID: | 3605 |
Gene Name: | interleukin 17A |
Gene Aliases: |
CTLA-8, CTLA8, IL-17, IL-17A, IL17 |
Location: |
Chr.6:52186387-52190638 on Build GRCh38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL17A | NM_002190.2 | NP_002181.1 | ||
| BC066251.1 | AAH66251.1 | |||
| BC066252.1 | AAH66252.1 | |||
| BC066253.1 | AAH66253.1 | |||
| BC067503.1 | AAH67503.1 | |||
| BC067504.1 | AAH67504.1 | |||
| BC067505.1 | AAH67505.1 | |||
| U32659.1 | AAC50341.1 | |||
| Z58820.1 | CAA91233.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522645 | Chr.6:52109491 - 52207217 on Build GRCh38 | Gain |
|
| nsv830662 | Chr.6:52064873 - 52264478 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map