Assay Details
Target Gene Details
Entrez Gene ID: | 6903 |
Gene Name: | tubulin folding cofactor C |
Gene Aliases: |
CFC |
Location: |
Chr.6:42744496-42746146 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBCC | NM_003192.2 | 1 | 395 | NP_003183.1 |
| AK312681.1 | 1 | 337 | ||
| BC017479.1 | 2 | 886 | AAH17479.1 | |
| BC020170.1 | 1 | 335 | AAH20170.1 | |
| BE795945.1 | 1 | 395 | ||
| BT007002.1 | 1 | 322 | AAP35648.1 | |
| U61234.1 | 1 | 345 | AAB17539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576145 | Chr.6:42728097 - 43012090 on Build GRCh38 | Gain |
|
| nsv830650 | Chr.6:42690493 - 42873400 on Build GRCh38 | Loss |
|
| nsv1017398 | Chr.6:42659705 - 43400998 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map