Assay Details
Target Gene Details
Entrez Gene ID: | 29785 |
Gene Name: | cytochrome P450 family 2 subfamily S member 1 |
Gene Aliases: |
CYPIIS1 |
Location: |
Chr.19:41193207-41207539 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP2S1 | NM_030622.7 | 3 | 464 | NP_085125.1 |
| AF335278.1 | 3 | 406 | AAK13498.1 | |
| AK027605.1 | 3 | 461 | BAB55227.1 | |
| AK296087.1 | ||||
| AY358603.1 | 3 | 449 | AAQ88966.1 | |
| BC033691.1 | 3 | 423 | AAH33691.1 | |
| BP284240.1 | 3 | 464 | ||
| BX422464.2 | 3 | 441 |
More Information
Set Membership: |
|

Genomic Map