Assay Details
Target Gene Details
Entrez Gene ID: | 1733 |
Gene Name: | deiodinase, iodothyronine type I |
Gene Aliases: |
5DI, TXDI1 |
Location: |
Chr.1:53894187-53911086 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIO1 | NM_000792.6 | NP_000783.2 | ||
| NM_001039715.2 | NP_001034804.1 | |||
| NM_001039716.2 | NP_001034805.1 | |||
| NM_001324316.1 | NP_001311245.1 | |||
| NM_213593.4 | NP_998758.1 | |||
| NR_136692.1 | ||||
| NR_136693.1 | ||||
| AK290780.1 | ||||
| AY560374.1 | AAT02480.1 | |||
| AY560375.1 | AAT02481.1 | |||
| AY560376.1 | AAT02482.1 | |||
| AY560377.1 | AAT02483.1 | |||
| AY560378.1 | AAT02484.1 | |||
| AY560379.1 | AAT02485.1 | |||
| AY560380.1 | AAT02486.1 | |||
| AY560381.1 | AAT02487.1 | |||
| AY560382.1 | AAT02488.1 | |||
| AY560383.1 | AAT02489.1 | |||
| BC017955.1 | AAH17955.2 | |||
| BC107170.2 | AAI07171.1 | |||
| BC107171.2 | AAI07172.1 | |||
| CD014030.1 | ||||
| FJ002243.1 | ||||
| FJ002244.1 | ||||
| FJ002245.1 | ||||
| FJ002247.1 | ||||
| FJ002248.1 | ||||
| FJ002249.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011262 | Chr.1:53542956 - 54106304 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map