Assay Details
Target Gene Details
Entrez Gene ID: | 5126 |
Gene Name: | proprotein convertase subtilisin/kexin type 2 |
Gene Aliases: |
NEC 2, NEC-2, NEC2, PC2, SPC2 |
Location: |
Chr.20:17226107-17484578 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCSK2 | NM_001201528.1 | NP_001188457.1 | ||
| NM_001201529.2 | NP_001188458.1 | |||
| NM_002594.4 | NP_002585.2 | |||
| BC040546.1 | AAH40546.1 | |||
| DA147868.1 | ||||
| DB173053.1 | ||||
| HY137524.1 | 1 | 65 |
More Information
Set Membership: |
|

Genomic Map