Assay Details
Target Gene Details
Entrez Gene ID: | 844 |
Gene Name: | calsequestrin 1 |
Gene Aliases: |
CASQ, PDIB1, VMCQA |
Location: |
Chr.1:160190495-160201886 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASQ1 | NM_001231.4 | 1 | 305 | NP_001222.3 |
| AK300713.1 | 1 | 213 | ||
| AK313250.1 | 1 | 208 | ||
| BC022289.1 | 1 | 191 | AAH22289.1 | |
| BX500008.1 | 1 | 225 | ||
| DA766051.1 | 1 | 305 | ||
| S73775.1 | 1 | 195 | AAB32063.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761950 | Chr.1:160158328 - 160193847 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map