Assay Details
Target Gene Details
Entrez Gene ID: | 54475 |
Gene Name: | notchless homolog 1 |
Gene Aliases: |
Nle |
Location: |
Chr.17:35128755-35142315 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLE1 | NM_001014445.2 | 10 | 1438 | NP_001014445.1 |
| NM_018096.4 | 11 | 1367 | NP_060566.2 | |
| XM_017024777.1 | 11 | 1385 | XP_016880266.1 | |
| AB209111.1 | 10 | 1438 | BAD92348.1 | |
| AJ005257.1 | 2 | 132 | CAA06444.1 | |
| AK001320.1 | 11 | 1356 | BAA91621.1 | |
| AK303253.1 | 11 | 1270 | ||
| AK304508.1 | 10 | 1229 | ||
| BC002884.2 | 10 | 1325 | AAH02884.2 | |
| BC012075.1 | 11 | 1340 | AAH12075.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2036 | Chr.17:35124777 - 35143140 on Build GRCh38 | Insertion |
|
| esv3892998 | Chr.17:35106468 - 35295755 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map