Assay Details
Target Gene Details
Entrez Gene ID: | 9715 |
Gene Name: | family with sequence similarity 131 member B |
Gene Aliases: |
- |
Location: |
Chr.7:143353400-143382304 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM131B | NM_001031690.2 | 7 | 2241 | NP_001026860.2 |
| NM_001278297.1 | 6 | 2147 | NP_001265226.1 | |
| NM_014690.4 | 7 | 2134 | NP_055505.3 | |
| XM_005250073.3 | 7 | 2455 | XP_005250130.1 | |
| XM_006716186.2 | 7 | 2233 | XP_006716249.1 | |
| AB018316.1 | 6 | 2029 | ||
| AK124557.1 | 7 | 2189 | ||
| AL832579.2 | 6 | 3722 | CAD89938.1 | |
| BC045611.1 | 7 | 2246 | AAH45611.2 | |
| BC050543.1 | 7 | 2109 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv949966 | Chr.7:143351408 - 143357207 on Build GRCh38 | Deletion |
|
| nsv517520 | Chr.7:143352880 - 143400007 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map