Assay Details
Target Gene Details
Entrez Gene ID: | 9536 |
Gene Name: | prostaglandin E synthase |
Gene Aliases: |
MGST-IV, MGST1-L1, MGST1L1, MPGES, PGES, PIG12, PP102, PP1294, TP53I12, mPGES-1 |
Location: |
Chr.9:129738336-129777579 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGES | NM_004878.4 | 3 | 1453 | NP_004869.1 |
| XM_011519210.2 | 4 | 1814 | XP_011517512.1 | |
| AF010316.1 | 3 | 1412 | AAC39534.1 | |
| AF027740.1 | AAB82299.1 | |||
| AF217965.1 | 4 | 1525 | AAG17208.1 | |
| AF217972.1 | 1 | 954 | AAG17215.1 | |
| AK127663.1 | 1 | 2595 | ||
| BC008280.1 | 3 | 1435 | AAH08280.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046477 | Chr.9:129608081 - 129997996 on Build GRCh38 | Gain |
|
| esv3621839 | Chr.9:129701704 - 129885823 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map