Assay Details
Target Gene Details
Entrez Gene ID: | 23001 |
Gene Name: | WD repeat and FYVE domain containing 3 |
Gene Aliases: |
ALFY, ZFYVE25 |
Location: |
Chr.4:84669537-84966391 on Build GRCh38 |
Assay Gene Location: | Within Exon 74 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDFY3 | NM_014991.4 | 68 | 11818 | NP_055806.2 |
| XM_005262858.4 | 70 | 11859 | XP_005262915.1 | |
| XM_011531757.2 | 69 | 11770 | XP_011530059.1 | |
| XM_011531759.2 | 68 | 11732 | XP_011530061.1 | |
| XM_011531760.2 | 68 | 11670 | XP_011530062.1 | |
| XM_011531761.2 | 69 | 11725 | XP_011530063.1 | |
| XM_011531762.2 | 68 | 11719 | XP_011530064.1 | |
| XM_011531763.2 | 67 | 11616 | XP_011530065.1 | |
| XM_011531764.2 | 68 | 11686 | XP_011530066.1 | |
| XM_011531765.2 | 65 | 11421 | XP_011530067.1 | |
| XM_017007906.1 | 71 | 11987 | XP_016863395.1 | |
| XM_017007907.1 | 71 | 11984 | XP_016863396.1 | |
| XM_017007908.1 | 70 | 11916 | XP_016863397.1 | |
| XM_017007909.1 | 56 | 9612 | XP_016863398.1 | |
| AB023210.2 | 33 | 5501 | BAA76837.2 | |
| BC150246.1 | 33 | 5522 | ||
| BX648077.1 | 25 | 4383 | ||
| BX648153.1 | 28 | 4711 |
More Information
Set Membership: |
|

Genomic Map