Assay Details
Target Gene Details
Entrez Gene ID: | 4092 |
Gene Name: | SMAD family member 7 |
Gene Aliases: |
CRCS3, MADH7, MADH8 |
Location: |
Chr.18:48919853-48950711 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMAD7 | NM_001190821.1 | 4 | 1394 | NP_001177750.1 |
| NM_001190822.1 | 4 | 602 | NP_001177751.1 | |
| NM_001190823.1 | 2 | 695 | NP_001177752.1 | |
| NM_005904.3 | 4 | 1397 | NP_005895.1 | |
| AF010193.1 | 4 | 1405 | AAB81246.1 | |
| AF015261.1 | 4 | 1110 | AAB81354.1 | |
| AK131394.1 | 2 | 637 | ||
| AK301535.1 | 4 | 602 | ||
| BC074818.2 | 4 | 1144 | AAH74818.2 | |
| BC074819.2 | 4 | 1144 | AAH74819.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833644 | Chr.18:48769390 - 48943545 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map