Assay Details
Target Gene Details
Entrez Gene ID: | 165215 |
Gene Name: | family with sequence similarity 171 member B |
Gene Aliases: |
KIAA1946 |
Location: |
Chr.2:186694062-186763787 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM171B | NM_177454.3 | NP_803237.3 | ||
| AF361495.1 | AAL57220.1 | |||
| AK091423.1 | BAC03660.1 | |||
| AK289737.1 | ||||
| BC060872.1 | AAH60872.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759107 | Chr.2:186468053 - 186715820 on Build GRCh38 | Gain |
|

Genomic Map