Assay Details
Target Gene Details
Entrez Gene ID: | 100127983 |
Gene Name: | chromosome 8 open reading frame 88 |
Gene Aliases: |
- |
Location: |
Chr.8:90958471-90985257 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C8orf88 | NM_001190972.1 | 6 | 734 | NP_001177901.1 |
| XM_017012939.1 | 6 | 660 | XP_016868428.1 | |
| BI562585.1 | 6 | 728 | ||
| BI829215.1 | 6 | 733 |
Target Gene Details
Entrez Gene ID: | 64168 |
Gene Name: | N-terminal EF-hand calcium binding protein 1 |
Gene Aliases: |
EFCBP1, STIP-1 |
Location: |
Chr.8:90791693-90959402 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NECAB1 | NM_022351.4 | 13 | 4535 | NP_071746.1 |
| AF414126.2 | 14 | 4624 | AAP33146.1 | |
| AK098356.1 | 1 | 293 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1028201 | Chr.8:90956299 - 90997736 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map