Assay Details
Target Gene Details
Entrez Gene ID: | 2925 |
Gene Name: | gastrin releasing peptide receptor |
Gene Aliases: |
BB2, BB2R |
Location: |
Chr.X:16123301-16153518 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRPR | NM_005314.2 | 1 | 591 | NP_005305.1 |
| AK313588.1 | 1 | 319 | ||
| BC074733.2 | AAH74733.1 | |||
| M73481.1 | 1 | 336 | AAA88050.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv436917 | Chr.X:16046179 - 16176281 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map