Assay Details
Target Gene Details
Entrez Gene ID: | 3062 |
Gene Name: | hypocretin receptor 2 |
Gene Aliases: |
OX2R |
Location: |
Chr.6:55174273-55284698 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HCRTR2 | NM_001526.3 | 1 | 466 | NP_001517.2 |
| XM_017010798.1 | 1 | 421 | XP_016866287.1 | |
| AF041245.1 | 1 | 510 | AAC39602.1 | |
| AK314279.1 | 1 | 418 | ||
| EF633107.1 | 2 | 671 | ||
| EF633108.1 | 3 | 770 | ||
| EF633109.1 | 2 | 735 | ||
| EF633110.1 | 1 | 1013 | ||
| KC812499.1 | 2 | 614 | ||
| KC812500.1 | 2 | 614 |
More Information
Set Membership: |
|

Genomic Map