Assay Details
Target Gene Details
Entrez Gene ID: | 30836 |
Gene Name: | deoxynucleotidyltransferase terminal interacting protein 2 |
Gene Aliases: |
ERBP, FCF2, HSU15552, LPTS-RP2, TdIF2 |
Location: |
Chr.1:93869458-93879206 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNTTIP2 | NM_014597.4 | NP_055412.2 | ||
| AB046574.1 | BAD08331.1 | |||
| AK123702.1 | 1 | 802 | ||
| AK222722.1 | BAD96442.1 | |||
| AK292379.1 | ||||
| AK302909.1 | ||||
| AK308808.1 | ||||
| AU134239.1 | ||||
| AY336729.1 | AAR02407.1 | |||
| AY394925.1 | AAQ95169.1 | |||
| BC047688.1 | AAH47688.1 | |||
| BC130622.1 | ||||
| U15552.1 | AAA50601.1 |
More Information
Set Membership: |
|

Genomic Map