Assay Details
Target Gene Details
Entrez Gene ID: | 100616248 |
Gene Name: | microRNA 4724 |
Gene Aliases: |
mir-4724 |
Location: |
Chr.17:31534883-31534971 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR4724 | NR_039877.1 |
Target Gene Details
Entrez Gene ID: | 84440 |
Gene Name: | RAB11 family interacting protein 4 |
Gene Aliases: |
RAB11-FIP4 |
Location: |
Chr.17:31391624-31538217 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB11FIP4 | NM_001303542.2 | 13 | 5023 | NP_001290471.2 |
| NM_032932.5 | 15 | 5316 | NP_116321.2 | |
| AB058724.1 | 15 | 5005 | BAB47450.1 | |
| AJ314646.2 | 15 | 5005 | CAC44535.1 | |
| AK055019.1 | 1 | 1914 | ||
| AY169244.1 | 15 | 5316 | AAO21970.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064728 | Chr.17:31511259 - 31545612 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map