Assay Details
Target Gene Details
Entrez Gene ID: | 147183 |
Gene Name: | keratin 25 |
Gene Aliases: |
ARWH3, KRT24IRS1, KRT25A |
Location: |
Chr.17:40748021-40755542 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT25 | NM_181534.3 | 1 | 169 | NP_853512.1 |
| XM_011524414.1 | 2 | 157 | XP_011522716.1 | |
| AJ564204.1 | 1 | 150 | CAD91904.1 | |
| AK129503.1 | 1 | 184 | BAC85169.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574995 | Chr.17:40722575 - 40767383 on Build GRCh38 | Loss |
|
| dgv5589n54 | Chr.17:40736437 - 40768780 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map