Assay Details
Target Gene Details
Entrez Gene ID: | 3162 |
Gene Name: | heme oxygenase 1 |
Gene Aliases: |
HMOX1D, HO-1, HSP32, bK286B10 |
Location: |
Chr.22:35381067-35394214 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMOX1 | NM_002133.2 | 5 | 1410 | NP_002124.1 |
| BC001491.2 | 5 | 1369 | AAH01491.1 | |
| CA306357.1 | 1 | 197 | ||
| M23042.1 | 1 | 337 | ||
| X06985.1 | 5 | 1383 | CAA30045.1 |
More Information
Set Membership: |
|

Genomic Map