Assay Details
Target Gene Details
Entrez Gene ID: | 2002 |
Gene Name: | ELK1, ETS transcription factor |
Gene Aliases: |
- |
Location: |
Chr.X:47635520-47650604 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELK1 | NM_001114123.2 | NP_001107595.1 | ||
| NM_001257168.1 | NP_001244097.1 | |||
| NM_005229.4 | NP_005220.2 | |||
| XM_017029339.1 | XP_016884828.1 | |||
| AB016193.1 | BAA36616.1 | |||
| AB209047.1 | 2 | 378 | BAD92284.1 | |
| AF000672.1 | AAD00862.1 | |||
| AK312984.1 | ||||
| BC048296.1 | AAH48296.1 | |||
| BC056150.1 | AAH56150.1 | |||
| CR542252.1 | CAG47048.1 | |||
| M25269.1 | AAA52384.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3573930 | Chr.X:47640398 - 47644194 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map