Assay Details
Target Gene Details
Entrez Gene ID: | 79870 |
Gene Name: | brain and acute leukemia, cytoplasmic |
Gene Aliases: |
- |
Location: |
Chr.8:103140693-103230305 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BAALC | NM_001024372.1 | NP_001019543.1 | ||
| NM_024812.2 | NP_079088.1 | |||
| AF371319.1 | AAL50515.1 | |||
| AK022077.1 | BAB13960.1 | |||
| AK093819.1 | ||||
| AK310816.1 | ||||
| BC011517.1 | AAH11517.1 | |||
| BC035038.2 | AAH35038.1 | |||
| BI548687.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831413 | Chr.8:103072913 - 103257561 on Build GRCh38 | Gain |
|
| esv21700 | Chr.8:103169891 - 103233301 on Build GRCh38 | Gain |
|
| nsv1017326 | Chr.8:103223965 - 103301007 on Build GRCh38 | Gain |
|

Genomic Map