Assay Details
Target Gene Details
Entrez Gene ID: | 9468 |
Gene Name: | phosphate cytidylyltransferase 1, choline, beta |
Gene Aliases: |
CCTB, CTB |
Location: |
Chr.X:24558087-24672862 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCYT1B | NM_001163264.1 | 6 | 832 | NP_001156736.1 |
| NM_001163265.1 | 6 | 889 | NP_001156737.1 | |
| NM_004845.4 | 6 | 889 | NP_004836.2 | |
| XM_017029977.1 | 7 | 714 | XP_016885466.1 | |
| AF052510.1 | 6 | 716 | AAC39754.1 | |
| AF148464.1 | 6 | 660 | AAD35088.1 | |
| AK226121.1 | 6 | 832 | ||
| AK296333.1 | 6 | 783 | ||
| AK315323.1 | 6 | 889 | ||
| BC045634.1 | 6 | 647 | AAH45634.2 |
More Information
Set Membership: |
|

Genomic Map