Assay Details
Target Gene Details
Entrez Gene ID: | 55 |
Gene Name: | acid phosphatase, prostate |
Gene Aliases: |
5'-NT, ACP-3, ACP3 |
Location: |
Chr.3:132317367-132368765 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACPP | NM_001099.4 | NP_001090.2 | ||
| NM_001134194.1 | NP_001127666.1 | |||
| NM_001292037.1 | NP_001278966.1 | |||
| XM_011512946.1 | XP_011511248.1 | |||
| XM_011512947.2 | XP_011511249.1 | |||
| AB102888.1 | BAD89417.1 | |||
| AK300540.1 | ||||
| BC007460.1 | AAH07460.1 | |||
| BC008493.1 | AAH08493.1 | |||
| BC016344.1 | AAH16344.1 | |||
| BC017877.1 | ||||
| DC383453.1 | ||||
| M24902.1 | AAA60022.1 | |||
| M34840.1 | AAA69694.1 | |||
| X52174.1 | CAA36422.1 | |||
| X53605.1 | CAA37673.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3597825 | Chr.3:132328496 - 132333897 on Build GRCh38 | Loss |
|
| esv2762346 | Chr.3:132162378 - 132364702 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map