Assay Details
Target Gene Details
Entrez Gene ID: | 5501 |
Gene Name: | protein phosphatase 1 catalytic subunit gamma |
Gene Aliases: |
PP-1G, PP1C, PPP1G |
Location: |
Chr.12:110708369-110742978 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1CC | NM_001244974.1 | NP_001231903.1 | ||
| NM_002710.3 | 7 | 1270 | NP_002701.1 | |
| XM_011538504.2 | XP_011536806.1 | |||
| XM_011538505.2 | XP_011536807.1 | |||
| AA781342.1 | ||||
| AK091841.1 | 1 | 995 | ||
| AK297882.1 | 7 | 988 | ||
| BC014073.2 | 7 | 1163 | AAH14073.1 | |
| L07395.1 | 7 | 987 | AAA19823.1 | |
| X74008.1 | 7 | 1153 | CAA52169.1 |
More Information
Set Membership: |
|

Genomic Map