Assay Details
Target Gene Details
Entrez Gene ID: | 6514 |
Gene Name: | solute carrier family 2 member 2 |
Gene Aliases: |
GLUT2 |
Location: |
Chr.3:170996341-171026979 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC2A2 | NM_000340.1 | 11 | 1854 | NP_000331.1 |
| NM_001278658.1 | 10 | 1591 | NP_001265587.1 | |
| NM_001278659.1 | 10 | 1729 | NP_001265588.1 | |
| XM_011513087.2 | 12 | 1601 | XP_011511389.1 | |
| XM_011513089.2 | 8 | 1142 | XP_011511391.1 | |
| AK290846.1 | 11 | 1597 | ||
| AK292741.1 | 11 | 1597 | ||
| AK298418.1 | 10 | 1355 | ||
| AK313622.1 | ||||
| BC060041.1 | 10 | 1474 | AAH60041.1 | |
| J03810.1 | 11 | 1583 | AAA59514.1 |
More Information
Set Membership: |
|

Genomic Map