Assay Details
Target Gene Details
Entrez Gene ID: | 645121 |
Gene Name: | cyclin I family member 2 |
Gene Aliases: |
- |
Location: |
Chr.5:132747402-132754403 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNI2 | NM_001039780.3 | NP_001034869.1 | ||
| NM_001287252.1 | NP_001274181.1 | |||
| NM_001287253.1 | NP_001274182.1 | |||
| XM_017009725.1 | XP_016865214.1 | |||
| AK131553.1 | BAD18687.1 | |||
| BC132837.1 | ||||
| BC136836.1 | ||||
| BC144416.1 | ||||
| BC144417.1 |
Target Gene Details
Entrez Gene ID: | 23176 |
Gene Name: | septin 8 |
Gene Aliases: |
SEP2 |
Location: |
Chr.5:132750817-132778216 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEPT8 | NM_001098811.1 | 10 | 2724 | NP_001092281.1 |
| XM_005271927.2 | 10 | 2723 | XP_005271984.1 | |
| XM_005271928.2 | 10 | 2720 | XP_005271985.1 | |
| XM_005271931.3 | 11 | 2807 | XP_005271988.1 | |
| XM_011543288.1 | 10 | 2746 | XP_011541590.1 | |
| XM_017009254.1 | 10 | 2717 | XP_016864743.1 | |
| XM_017009255.1 | 10 | 2790 | XP_016864744.1 | |
| XM_017009256.1 | 10 | 2787 | XP_016864745.1 | |
| XM_017009257.1 | 10 | 2740 | XP_016864746.1 | |
| AF440762.1 | 10 | 2724 | AAO13879.1 | |
| BU739074.1 | 1 | 184 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528687 | Chr.5:132750285 - 132779155 on Build GRCh38 | Gain |
|
| esv3606809 | Chr.5:132749939 - 132764024 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map