Assay Details
Target Gene Details
Entrez Gene ID: | 100505996 |
Gene Name: | long intergenic non-protein coding RNA 403 |
Gene Aliases: |
- |
Location: |
Chr.13:111972310-112108015 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00403 | NR_120392.1 | |||
| AK091058.1 | ||||
| DA195709.1 |
Target Gene Details
Entrez Gene ID: | 6656 |
Gene Name: | SRY-box 1 |
Gene Aliases: |
- |
Location: |
Chr.13:112067599-112071706 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOX1 | NM_005986.2 | 1 | 1521 | NP_005977.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1069775 | Chr.13:112067785 - 112069086 on Build GRCh38 | Deletion |
|
| dgv351n67 | Chr.13:112033868 - 112116516 on Build GRCh38 | Gain |
|
| nsv563189 | Chr.13:111979751 - 112146300 on Build GRCh38 | Loss |
|
| nsv832722 | Chr.13:112055927 - 112247292 on Build GRCh38 | Loss |
|
| dgv352n67 | Chr.13:112064263 - 112071929 on Build GRCh38 | Gain |
|
| nsv826816 | Chr.13:112048745 - 112080890 on Build GRCh38 | Gain |
|
| nsv826815 | Chr.13:112039819 - 112098509 on Build GRCh38 | Loss |
|
| nsv826807 | Chr.13:112034289 - 112083084 on Build GRCh38 | Gain |
|
| nsv826813 | Chr.13:112038383 - 112141293 on Build GRCh38 | Gain |
|
| nsv1112639 | Chr.13:112067786 - 112069086 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map