Assay Details
Target Gene Details
Entrez Gene ID: | 80834 |
Gene Name: | taste 1 receptor member 2 |
Gene Aliases: |
GPR71, T1R2, TR2 |
Location: |
Chr.1:18839599-18859661 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAS1R2 | NM_152232.2 | NP_689418.2 |
More Information
Set Membership: |
|

Genomic Map