Assay Details
Target Gene Details
Entrez Gene ID: | 6567 |
Gene Name: | solute carrier family 16 member 2 |
Gene Aliases: |
AHDS, DXS128, DXS128E, MCT 7, MCT 8, MCT7, MCT8, MRX22, XPCT |
Location: |
Chr.X:74421493-74533929 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC16A2 | NM_006517.4 | 1 | 476 | NP_006508.2 |
| AB085789.1 | 1 | 418 | BAC76827.1 | |
| CN342861.1 | 1 | 477 | ||
| U05315.1 | 1 | 508 | AAB60374.1 |
More Information
Set Membership: |
|

Genomic Map