Assay Details
Target Gene Details
Entrez Gene ID: | 5516 |
Gene Name: | protein phosphatase 2 catalytic subunit beta |
Gene Aliases: |
PP2Abeta, PP2CB |
Location: |
Chr.8:30785610-30812836 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2CB | NM_001009552.1 | NP_001009552.1 | ||
| AF444006.1 | AAL35904.1 | |||
| AK091875.1 | ||||
| BC012022.1 | AAH12022.1 | |||
| BT019526.1 | AAV38333.1 | |||
| CR541747.1 | CAG46547.1 | |||
| J03805.1 | AAB38020.1 | |||
| X12656.1 | CAA31183.1 |
More Information
Set Membership: |
|

Genomic Map