Assay Details
Target Gene Details
Entrez Gene ID: | 11189 |
Gene Name: | CUGBP, Elav-like family member 3 |
Gene Aliases: |
BRUNOL1, CAGH4, ERDA4, ETR-1, TNRC4 |
Location: |
Chr.1:151700058-151716821 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952176 | Chr.1:151701725 - 151717424 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map