Assay Details
Target Gene Details
Entrez Gene ID: | 92346 |
Gene Name: | chromosome 1 open reading frame 105 |
Gene Aliases: |
- |
Location: |
Chr.1:172418560-172468829 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf105 | NM_139240.3 | NP_640333.3 | ||
| XM_005245608.2 | XP_005245665.1 | |||
| XM_005245610.3 | XP_005245667.1 | |||
| XM_006711630.2 | XP_006711693.1 | |||
| XM_011510152.2 | XP_011508454.1 | |||
| XM_011510153.2 | XP_011508455.1 | |||
| XM_011510154.2 | XP_011508456.1 | |||
| XM_011510155.2 | XP_011508457.1 | |||
| XM_011510156.2 | XP_011508458.1 | |||
| XM_017002808.1 | XP_016858297.1 | |||
| AL035295.1 | CAA22895.1 | |||
| BC038410.1 | AAH38410.1 |
More Information
Set Membership: |
|

Genomic Map