Assay Details
Target Gene Details
Entrez Gene ID: | 51605 |
Gene Name: | tRNA methyltransferase 6 |
Gene Aliases: |
CGI-09, GCD10, Gcd10p |
Location: |
Chr.20:5937832-5950558 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRMT6 | NM_001281467.1 | 10 | 1969 | NP_001268396.1 |
| NM_015939.4 | 11 | 2079 | NP_057023.2 | |
| XM_017027875.1 | 11 | 2175 | XP_016883364.1 | |
| AB032979.1 | 9 | 2705 | BAA86467.1 | |
| AI291549.1 | 1 | 275 | ||
| AK000613.1 | 11 | 2005 | ||
| BC001262.1 | 11 | 1998 | AAH01262.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833905 | Chr.20:5774308 - 5941219 on Build GRCh38 | Gain |
|
| nsv819052 | Chr.20:5931265 - 5938364 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map