Assay Details
Target Gene Details
Entrez Gene ID: | 343406 |
Gene Name: | olfactory receptor family 10 subfamily R member 2 |
Gene Aliases: |
OR1-8, OR10R2Q |
Location: |
Chr.1:158479878-158480885 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR10R2 | NM_001004472.1 | 1 | 605 | NP_001004472.1 |
More Information
Set Membership: |
|

Genomic Map