Assay Details
Target Gene Details
Entrez Gene ID: | 254028 |
Gene Name: | uncharacterized LOC254028 |
Gene Aliases: |
- |
Location: |
Chr.14:20588224-20607221 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC254028 | NR_122043.1 | |||
| BI830942.1 | ||||
| DB034207.1 | ||||
| DB053670.1 |
Target Gene Details
Entrez Gene ID: | 122651 |
Gene Name: | ribonuclease A family member 11 (inactive) |
Gene Aliases: |
C14orf6, HEL-S-84p, RAJ1 |
Location: |
Chr.14:20582892-20590258 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASE11 | NM_145250.3 | 1 | 50 | NP_660293.1 |
| AY358794.1 | 1 | 331 | AAQ89154.1 | |
| BX161458.1 | CAD61920.1 | |||
| CB995247.1 | 1 | 76 |
Target Gene Details
Entrez Gene ID: | 493901 |
Gene Name: | ribonuclease A family member 12 (inactive) |
Gene Aliases: |
HEL-S-85p, RAI1 |
Location: |
Chr.14:20590080-20590823 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASE12 | NM_001024822.2 | 1 | 615 | NP_001019993.1 |
| AW274139.1 | 1 | 134 | ||
| BC130347.1 | ||||
| BC130349.1 | ||||
| FJ237362.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23438 | Chr.14:20332606 - 20789719 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map