Assay Details
Target Gene Details
Entrez Gene ID: | 3916 |
Gene Name: | lysosomal associated membrane protein 1 |
Gene Aliases: |
CD107a, LAMPA, LGP120 |
Location: |
Chr.13:113297154-113323431 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAMP1 | NM_005561.3 | NP_005552.3 | ||
| XM_011537494.1 | XP_011535796.1 | |||
| AB209319.1 | BAD92556.1 | |||
| AK092398.1 | ||||
| AK301584.1 | ||||
| BC006345.2 | AAH06345.2 | |||
| BC007845.2 | AAH07845.2 | |||
| BC021288.2 | AAH21288.1 | |||
| BC093044.1 | AAH93044.1 | |||
| BU849686.1 | ||||
| J03263.1 | AAA59524.1 | |||
| J04182.1 | AAA60382.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3633598 | Chr.13:113310386 - 113324953 on Build GRCh38 | Loss |
|
| nsv1048814 | Chr.13:113081728 - 113611496 on Build GRCh38 | Gain |
|
| nsv509521 | Chr.13:113308960 - 113394480 on Build GRCh38 | Insertion |
|
| nsv563316 | Chr.13:113261171 - 113347907 on Build GRCh38 | Loss |
|
| nsv563317 | Chr.13:113292168 - 113369086 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map