Assay Details
Target Gene Details
Entrez Gene ID: | 26033 |
Gene Name: | attractin like 1 |
Gene Aliases: |
ALP, bA338L11.1, bA454H24.1 |
Location: |
Chr.10:115093365-115948999 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATRNL1 | NM_001276282.3 | 1 | 593 | NP_001263211.1 |
| NM_207303.4 | 1 | 593 | NP_997186.1 | |
| NR_074088.2 | ||||
| XM_011539587.1 | 1 | 588 | XP_011537889.1 | |
| XM_011539588.2 | XP_011537890.1 | |||
| XM_011539589.2 | 1 | 588 | XP_011537891.1 | |
| XM_017016035.1 | 1 | 588 | XP_016871524.1 | |
| XM_017016036.1 | 1 | 588 | XP_016871525.1 | |
| AK127277.1 | BAC86914.1 | |||
| AY442317.1 | 1 | 207 | AAR14297.1 | |
| BC035157.1 | 1 | 598 | AAH35157.1 | |
| BC047716.1 | 1 | 538 | AAH47716.1 | |
| BC139916.1 | 1 | 582 | ||
| BC139923.1 | 1 | 582 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038050 | Chr.10:114555350 - 115135954 on Build GRCh38 | Gain |
|
| esv3624624 | Chr.10:114876531 - 115123608 on Build GRCh38 | Gain |
|
| nsv831998 | Chr.10:115063088 - 115209866 on Build GRCh38 | Loss |
|
| esv3577749 | Chr.10:114884410 - 115119719 on Build GRCh38 | Gain |
|
| dgv977n100 | Chr.10:114883336 - 115119719 on Build GRCh38 | Gain |
|
| nsv831997 | Chr.10:114929793 - 115095941 on Build GRCh38 | Gain |
|
| esv3624626 | Chr.10:114923110 - 115094807 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map