Assay Details
Target Gene Details
Entrez Gene ID: | 64860 |
Gene Name: | armadillo repeat containing, X-linked 5 |
Gene Aliases: |
GASP5 |
Location: |
Chr.X:102599168-102604159 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARMCX5 | NM_001168478.1 | 4 | 1287 | NP_001161950.1 |
| NM_001168479.1 | 6 | 1403 | NP_001161951.1 | |
| NM_001168480.1 | 6 | 1397 | NP_001161952.1 | |
| NM_001168482.1 | 3 | 1186 | NP_001161954.1 | |
| NM_001168485.1 | 5 | 1284 | NP_001161957.1 | |
| NM_022838.3 | 3 | 1174 | NP_073749.2 | |
| AK023031.1 | 2 | 1151 | BAB14367.1 | |
| AK096692.1 | 6 | 1397 | ||
| BC058904.1 | 3 | 1129 | AAH58904.1 | |
| BC064983.1 | 3 | 1442 | AAH64983.1 | |
| CR749476.1 | 1 | 3139 | CAH18306.1 |
Target Gene Details
Entrez Gene ID: | 100528062 |
Gene Name: | ARMCX5-GPRASP2 readthrough |
Gene Aliases: |
- |
Location: |
Chr.X:102599348-102717733 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARMCX5-GPRASP2 | NM_001199818.1 | NP_001186747.1 | ||
| BG392679.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526716 | Chr.X:102582805 - 102934382 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map