Assay Details
Target Gene Details
Entrez Gene ID: | 781 |
Gene Name: | calcium voltage-gated channel auxiliary subunit alpha2delta 1 |
Gene Aliases: |
CACNA2, CACNL2A, CCHL2A, LINC01112, lncRNA-N3 |
Location: |
Chr.7:81946444-82443921 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNA2D1 | NM_000722.3 | NP_000713.2 | ||
| XM_005250570.2 | XP_005250627.1 | |||
| XM_005250572.2 | XP_005250629.1 | |||
| XM_005250573.2 | XP_005250630.1 | |||
| XM_005250574.2 | XP_005250631.1 | |||
| XM_006716118.2 | XP_006716181.1 | |||
| XM_006716119.3 | XP_006716182.1 | |||
| XM_006716120.3 | XP_006716183.1 | |||
| XM_011516570.2 | XP_011514872.1 | |||
| XM_011516571.2 | XP_011514873.1 | |||
| XM_011516572.2 | XP_011514874.1 | |||
| XM_017012588.1 | XP_016868077.1 | |||
| BC117468.1 | ||||
| BC117470.1 | ||||
| BG055272.1 | ||||
| M76559.1 | AAA51903.1 |
Target Gene Details
Entrez Gene ID: | 101927356 |
Gene Name: | uncharacterized LOC101927356 |
Gene Aliases: |
- |
Location: |
Chr.7:82009177-82029955 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927356 | NR_110076.1 | |||
| NR_110077.1 | ||||
| AK055932.1 | ||||
| BM468380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026154 | Chr.7:81952009 - 82027249 on Build GRCh38 | Gain |
|
| nsv1019753 | Chr.7:81692852 - 82052256 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map