Assay Details
Target Gene Details
Entrez Gene ID: | 4643 |
Gene Name: | myosin IE |
Gene Aliases: |
FSGS6, HuncM-IC, MYO1C |
Location: |
Chr.15:59135969-59372872 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 25 - Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYO1E | NM_004998.3 | NP_004989.2 | ||
| AB290177.1 | ||||
| AK291688.1 | ||||
| BC098392.1 | AAH98392.1 | |||
| BX489024.1 | ||||
| U14391.1 | AAA62667.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527307 | Chr.15:58811129 - 59726288 on Build GRCh38 | Gain |
|
| nsv833025 | Chr.15:59043949 - 59229977 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map